HPLC Analysis of Flavanone Enantiomers on Astec® CYCLOBOND® I 2000 DMP, Ethanol:Hexane Mobile Phase
HPLC Analysis of Flavanone Enantiomers on Astec® CYCLOBOND® I 2000 DMP, Ethanol:Hexane Mobile Phase
Conditions
column CYCLOBOND I 2000 DMP, 25 cm x 4.6 mm I.D., 5 µm particles (20724AST)
column temp. 25 °C
mobile phase [A] ethanol; [B] hexane; (20:80, A:B)
flow rate 1 mL/min
sample 1 mg/mL in methanol
injection 1 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Chiral Applications, Preservatives, Food & Beverage, Antioxidants, All Food & Beverage Applications
Featured Industry Vitamins, Nutraceuticals, and Natural Products
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CYCLOBOND is a registered trademark of Sigma-Aldrich Co. LLC