HPLC Analysis of Flavones on Discovery® HS PEG
HPLC Analysis of Flavones on Discovery® HS PEG
Conditions
column Discovery HS PEG, 15 cm x 4.6 mm I.D., 5 µm particles (567416-U)
column temp. 30 °C
mobile phase [A] 0.1% formic acid in water; [B] 0.1% formic acid in methanol; (45:55, A:B)
flow rate 1.0 mL/min
injection 10 µL, each compound 50 µg/mL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Flavors, Fragrances
Featured Industry Food and Beverages
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC