HPLC Analysis of Flavonoids on Ascentis® RP-Amide
HPLC Analysis of Flavonoids on Ascentis® RP-Amide
Conditions
column Ascentis RP-Amide, 15 cm x 4.6 mm I.D., 5 µm particles (565324-U)
column temp. 30 °C
mobile phase [A] 0.085% orthophosphoric acid; [B] acetonitrile;
flow rate 1.0 mL/min
sample as indicated in mobile phase A
injection 10 µL
detector UV, 280 nm
Description
Categories Analytical Chromatography, Flavors, Fragrances
Featured Industry Vitamins, Nutraceuticals, and Natural Products