HPLC Analysis of Fluorinated Corticosteroids on Ascentis® C18
HPLC Analysis of Fluorinated Corticosteroids on Ascentis® C18
Conditions
column Ascentis C18, 5 cm x 4.6 mm I.D., 5 µm particles (581323-U)
column temp. 30 °C
mobile phase [A] water; [B] acetonitrile;
flow rate 1.0 mL/min
sample as indicated in acetonitrile
injection 5 µL
detector UV, 240 nm
Description
Categories Analytical Chromatography, Clinical, Steroids, steroids