HPLC Analysis of Fluoxetine Enantiomers on Astec® CHIROBIOTIC® V2
HPLC Analysis of Fluoxetine Enantiomers on Astec® CHIROBIOTIC® V2
Conditions
column CHIROBIOTIC V2, 25 cm x 4.6 mm I.D., 5 µm particles (15024AST)
column temp. 25 °C
mobile phase 15 mM ammonium acetate in methanol
flow rate 1 mL/min
sample 1 mg/mL in methanol
injection 5 µL
detector UV, 230 nm
Description
Categories Analytical Chromatography, Chiral Applications, Pharmaceutical (small molecule), Antidepressants, Clinical, Forensic & Toxicology, All Pharmaceutical Applications, Therapeutic Drug Monitoring