HPLC Analysis of Hydrophobic Bases on Ascentis® RP-Amide
HPLC Analysis of Hydrophobic Bases on Ascentis® RP-Amide
Conditions
column Ascentis RP-Amide, 15 cm x 4.6 mm I.D., 5 µm particles (565324-U)
column temp. 35 °C
mobile phase [A] 25 mM ammonium phosphate, pH 7; [B] methanol; (35:65, A:B)
flow rate 1.2 mL/min
sample N05158
injection 10 µL
detector UV, 230 nm
Description
Categories Analytical Chromatography, Organic Bases