HPLC Analysis of Hydroxymetoprolol, Metoprolol and Alprenolol on Discovery® HS F5
HPLC Analysis of Hydroxymetoprolol, Metoprolol and Alprenolol on Discovery® HS F5
Conditions
column Discovery HS F5, 5 cm x 2.1 mm I.D., 3 µm particles (567500-U)
column temp. 60 °C
mobile phase 5 mM ammonium acetate in 10:90 water:acetonitrile
flow rate 0.2 mL/min
sample as indicated (in mobile phase)
injection 10 µL
detector (+) ESI
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Antihypertensives
Featured Industry Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC