HPLC Analysis of Insulin, Bovine on Discovery® C8
HPLC Analysis of Insulin, Bovine on Discovery® C8
Conditions
column Discovery C8, 5 cm x 4.6 mm I.D., 5 µm particles (59353-U)
column temp. 35 °C
mobile phase 50 mM sodium phosphate, monobasic, 50 mM sodium sulfate, pH 3.0 (w/phosphoric acid), (A) +5% acetonitrile, (B) +50% acetonitrile
flow rate 1 mL/min
detector UV, 220 nm
Description
Categories Analytical Chromatography, Hormones
Featured Industry Life Science and Biopharma
Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC