HPLC Analysis of Insulin on SRT®-C SEC-300
HPLC Analysis of Insulin on SRT®-C SEC-300
Conditions
column SRT-C SEC-300, 30 cm x 7.8 mm I.D., 5 µm particles (Z777110)
column temp. ambient
mobile phase 150 mM sodium phosphate, pH 7
flow rate 1.0 mL/min
sample insulin
injection 10 µL
detector UV, 214 nm
Description
Categories Analytical Chromatography, Peptides
Featured Industry Life Science and Biopharma
Legal Information SRT is a registered trademark of Sepax Technologies