HPLC Analysis of Jacobsen Catalyst Enantiomers on Astec® CYCLOBOND® I 2000 HP-RSP (TEA mobile phase)
HPLC Analysis of Jacobsen Catalyst Enantiomers on Astec® CYCLOBOND® I 2000 HP-RSP (TEA mobile phase)
Conditions
column CYCLOBOND I 2000 HP-RSP, 25 cm x 4.6 mm I.D., 5 µm particles (24024AST)
column temp. 25 °C
mobile phase [A] acetonitrile; [B] methanol; [C] acetic acid; [D] triethylamine; (95:5:0.2:0.1, A:B:C:D)
flow rate 1.5 mL/min
sample 5 mg/mL in methanol
injection 2 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Chiral Applications, Catalysts, Organic Chemicals, Chemicals and Industrial Polymers
Featured Industry Chemicals and Industrial Polymers
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CYCLOBOND is a registered trademark of Sigma-Aldrich Co. LLC