HPLC Analysis of Lactic Acid Enantiomers on Astec® CLC-L, Trace Level Detection
HPLC Analysis of Lactic Acid Enantiomers on Astec® CLC-L, Trace Level Detection
Conditions
column Astec CLC-L, 15 cm x 4.6 mm I.D., 5 µm particles (53123AST)
column temp. 25 °C
mobile phase 5 mM copper sulfate
flow rate 1 mL/min
sample 5 mg/mL in water
injection 10 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Chiral Applications, Food & Beverage, Organic Acids, Chemicals and Industrial Polymers, All Food & Beverage Applications
Featured Industry Chemicals and Industrial Polymers
Food and Beverages
Vitamins, Nutraceuticals, and Natural Products
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC