HPLC Analysis of LMW Heparin-Dalteparin and Enoxaparin Analysis on Zenix®-100
HPLC Analysis of LMW Heparin-Dalteparin and Enoxaparin Analysis on Zenix®-100
Conditions
column Zenix SEC-100, 30 cm x 7.8 mm I.D., 3 µm particles, 100 Ã… (Z777008)
column temp. 35 °C
mobile phase Sodium sulfate, 2.84%, pH 5.0
flow rate 0.5 mL/min
sample 10 mg/mL Dalteparin in water, pH 6.0, MW 3,000 - 8,000 Da (1st chromatogram)
sample 10 mg/mL Enoxaparin in mobile phase, MW 3,000 - 8,000 Da (2nd chroatogram)
injection 25 µL
detector UV 234 nm and RI
Description
Analysis Note High resolution separations of low molecular weight heparin-daletparin and enoxaparin can be achived using Zenix SEC-100 columns.
Categories Analytical Chromatography, Anticoagulants
Featured Industry Life Science and Biopharma
Legal Information Zenix is a registered trademark of Sepax Technologies