Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of mAb on Antibodix® WCX-NP10, Competitive Comparison
HPLC Analysis of mAb on Antibodix® WCX-NP10, Competitive Comparison
Conditions
column Antibodix WCX-NP10, 25 cm x 4.6 mm I.D., 10 µm particles (Z777272)
column temp. ambient
mobile phase (A) 10 mM phosphate buffer, pH 7.5; (B) A + 100 mM NaCl
gradient 15 to 55% B in 30 min
flow rate 0.8 ml/min
sample monoclonal antibody
injection 5 µl
detector UV, 214 nm
Description
Categories Analytical Chromatography, Antibiotics, Antimicrobials
Featured Industry Life Science and Biopharma
Pharmaceutical (small molecule)
Legal Information Antibodix is a registered trademark of Sepax Technologies