HPLC Analysis of Methylphenidate (Ritalin) Enantiomers on Astec® CHIROBIOTIC® T2
HPLC Analysis of Methylphenidate (Ritalin) Enantiomers on Astec® CHIROBIOTIC® T2
Conditions
column CHIROBIOTIC T2, 25 cm x 4.6 mm I.D., 5 µm particles (16024AST)
column temp. 25 °C
mobile phase 15 mM ammonium formate in methanol
flow rate 1 mL/min
sample methylphenidate, 1 mg/mL in mobile phase
injection 5 µL
detector UV, 220 nm
Description
Categories Analytical Chromatography, Chiral Applications, Clinical, AD/HD Drugs, Clinical, Forensic & Toxicology, ADHD drugs, Pharmaceutical (small molecule), Therapeutic Drug Monitoring, Drugs of Abuse, All Pharmaceutical Applications
Featured Industry Clinical
Pharmaceutical (small molecule)