HPLC Analysis of Metoprolol Enantiomers on Astec® CHIROBIOTIC® V
HPLC Analysis of Metoprolol Enantiomers on Astec® CHIROBIOTIC® V
Conditions
column CHIROBIOTIC V, 25 cm x 4.6 mm I.D., 5 µm particles (11024AST)
column temp. 20 °C
mobile phase [A] methanol; [B] acetic acid; [C] triethylamine; (99.8:0.1:0.1, A:B:C)
flow rate 0.5 mL/min
sample 1 mg/mL in methanol
injection 2 µL
detector UV, 225 nm
Description
Categories Analytical Chromatography, Chiral Applications, Antihypertensives, Pharmaceutical (small molecule), All Pharmaceutical Applications