HPLC Analysis of Mianserin Enantiomers on Astec® Cellulose DMP, POM Conditions
HPLC Analysis of Mianserin Enantiomers on Astec® Cellulose DMP, POM Conditions
Conditions
column Astec Cellulose DMP, 15 cm x 4.6 mm I.D., 5 µm particles (51098AST)
column temp. 25 °C
mobile phase [A] methanol; [B] DEA; (100:0.1, A:B)
flow rate 0.5 mL/min
sample 2 mg/mL in mobile phase
inj. temp. 2 µL
detector UV, 230 nm
Description
Categories Analytical Chromatography, Antidepressants
Featured Industry Pharmaceutical (small molecule)
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC