HPLC Analysis of Miconazole Enantiomers on Astec® CYCLOBOND® I 2000 HP-RSP
HPLC Analysis of Miconazole Enantiomers on Astec® CYCLOBOND® I 2000 HP-RSP
Conditions
column CYCLOBOND I 2000 HP-RSP, 25 cm x 4.6 mm I.D., 5 µm particles (24024AST)
column temp. 25 °C
mobile phase [A] acetonitrile; [B] 20 mM ammonium acetate, pH 4.0; (25:75, A:B)
flow rate 0.6 mL/min
sample miconazole, 1 mg/mL in acetonitrile:water (50:50)
injection 5 µL
detector UV, 230 nm
Description
Categories Analytical Chromatography, Antibiotics, Antimicrobials
Featured Industry Pharmaceutical (small molecule)
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CYCLOBOND is a registered trademark of Sigma-Aldrich Co. LLC