HPLC Analysis of Monoclonal Antibody Heavy and Light Chains on Zenix® SEC-300
HPLC Analysis of Monoclonal Antibody Heavy and Light Chains on Zenix® SEC-300
Conditions
column Zenix SEC-300, 30 cm x 7.8 mm I.D., 3 µm particles (Z777033)
mobile phase 0.1% TFA, 0.1% formic acid, 20% acetonitrile
flow rate 0.5 mL/min
sample Monoclonal antibody (1 mg/mL) reduced with 20 mM DTT
injection 20 µL
detector UV, 280nm
Description
Categories Analytical Chromatography, Peptides
Featured Industry Life Science and Biopharma
Legal Information Zenix is a registered trademark of Sepax Technologies