HPLC Analysis of Morphine and Glucuronide Metabolites on Discovery® HS F5
HPLC Analysis of Morphine and Glucuronide Metabolites on Discovery® HS F5
Conditions
column Discovery HS F5, 5 cm x 4.6 mm I.D., 5 µm particles (567513-U)
column temp. 40 °C
mobile phase [A] 10 mM ammonium acetate, pH 6.8 (unadjusted); [B] acetonitrile
gradient 0% B for 1 min, to 30% B in 4 min, held 3 min
flow rate 1 mL/min
sample 50 µg/mL in final mobile phase
injection 10 µL
detector UV, 280 nm
Description
Categories Analytical Chromatography, Clinical, drugs of abuse, Drugs of Abuse
Featured Industry Clinical
Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC