HPLC Analysis of N-Acetylmethionine Enantiomers on Astec® (R,R) P-CAP™
HPLC Analysis of N-Acetylmethionine Enantiomers on Astec® (R,R) P-CAP™
Conditions
column Astec (R,R) P-CAP, 25 cm x 4.6 mm I.D., 5 µm particles (31024AST)
column temp. 25 °C
mobile phase 20 mM ammonium acetate in: (A) acetonitrile; (B) methanol; (70:30, A:B)
flow rate 1 mL/min
sample 1 mg/mL in methanol
injection 1 µL
detector UV, 230 nm
Description
Categories Analytical Chromatography, Chiral Applications, Amino Acids, Life Science & Biopharma, All Biochemical Applications, Native Amino Acids, All Amino Acids
Featured Industry Life Science and Biopharma
Legal Information P-CAP is a trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC