HPLC Analysis of Nicardipine Enantiomers on Astec® CHIROBIOTIC® V2
HPLC Analysis of Nicardipine Enantiomers on Astec® CHIROBIOTIC® V2
Conditions
column CHIROBIOTIC V2, 25 cm x 4.6 mm I.D., 5 µm particles (15024AST)
column temp. 30 °C
mobile phase [A] methanol; [B] 20 mM ammonium acetate, pH 4.1; (95:5, A:B)
flow rate 1 mL/min
sample 1 mg/mL in methanol
injection 5 µL
detector UV, 230 nm
Description
Categories Analytical Chromatography, Chiral Applications, Pharmaceutical (small molecule), Antihypertensives, All Pharmaceutical Applications