HPLC Analysis of Norphenylephrine Enantiomers on Astec® CYCLOBOND® I 2000 AC (pH 4.1)
HPLC Analysis of Norphenylephrine Enantiomers on Astec® CYCLOBOND® I 2000 AC (pH 4.1)
Conditions
column CYCLOBOND I 2000 AC, 25 cm x 4.6 mm I.D., 5 µm particles (20124AST)
column temp. 25 °C
mobile phase [A] 20 mM ammonium acetate, pH 4.1; [B] methanol; (93:7, A:B)
flow rate 0.5 mL/min
sample 1 mg/mL in methanol
injection 1 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Chiral Applications, Pharmaceutical (small molecule), Catecholamines, All Pharmaceutical Applications
Featured Industry Life Science and Biopharma
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CYCLOBOND is a registered trademark of Sigma-Aldrich Co. LLC