HPLC Analysis of Nucleosides on Ascentis® Phenyl
HPLC Analysis of Nucleosides on Ascentis® Phenyl
Conditions
column Ascentis Phenyl, 15 cm x 2.1 mm I.D., 5 µm particles (581613-U)
column temp. 35 °C
mobile phase 10 mM ammonium formate (pH 3.0 with formic acid)
flow rate 0.2 mL/min
sample as indicated, in mobile phase
injection 1 µL
detector UV, 270 nm; 750 psi back pressure regulator on outlet of flow cell
Description
Categories Analytical Chromatography, Nucleic Acids