HPLC Analysis of Oligosaccharides from Dark Corn Syrup on SUPELCOGEL™ Ag
HPLC Analysis of Oligosaccharides from Dark Corn Syrup on SUPELCOGEL™ Ag
Conditions
column SUPELCOGEL Ag, 30 cm x 7.8 mm I.D. (59315)
column temp. 85 °C
mobile phase water
flow rate 0.5 mL/min
injection 10 µLof 1g syrup/10 mL DI water, filtered (0.20 µm filter)
detector RI
Description
Categories Analytical Chromatography, Carbohydrates, Polysaccharides
Featured Industry Food and Beverages
Legal Information SUPELCOGEL is a trademark of Sigma-Aldrich Co. LLC