HPLC Analysis of Organic Acids in Riesling Wine on SeQuant ZIC-HILIC
HPLC Analysis of Organic Acids in Riesling Wine on SeQuant ZIC-HILIC
Conditions
column SeQuant ZIC-HILIC (5 µm, 200 Ã…) PEEK, 150x2.1 mm (1.50454.0001)
column temp. 30 °C
mobile phase [A] acetonitrile; [B] potassium phosphate; (75:25, A:B)
flow rate 0.4 mL/min
sample Riesling wine or 10 ppm mix of standards
injection 5 µL
detector 200 nm
detector Shimadzu LC-10Vp equipped with 2.5µL semi-micro flow-cell