HPLC Analysis of Organophosphorus Pesticides on Discovery® Cyano
HPLC Analysis of Organophosphorus Pesticides on Discovery® Cyano
Conditions
column Discovery Cyano, 15 cm x 4.6 mm I.D., 5 µm particles (59356-U)
column temp. 20 °C
mobile phase (A) water: (B) acetonitrile, (75:25, A:B)
flow rate 2 mL/min
injection 1 µL
detector UV, 214 nm
Description
Categories Analytical Chromatography, Food & Beverage Analysis, Environmental Analysis, Pesticides & Herbicides, Pesticide and Metabolite Residues, Organics, Applications (Chromatograms), Chromatograms, HPLC Chromatograms
Featured Industry Environmental
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC