HPLC Analysis of Oxprenolol Enantiomers on Astec® CHIROBIOTIC® T2
HPLC Analysis of Oxprenolol Enantiomers on Astec® CHIROBIOTIC® T2
Conditions
column CHIROBIOTIC T2, 25 cm x 4.6 mm I.D., 5 µm particles (16024AST)
column temp. 25 °C
mobile phase 5 mM ammonium formate in methanol
flow rate 1 mL/min
sample 1 mg/mL in methanol
injection 5 µL
detector UV, 220 nm
Description
Categories Analytical Chromatography, Chiral Applications, Pharmaceutical (small molecule), Antihypertensives, All Pharmaceutical Applications