HPLC Analysis of Paraquat and Diquat on Ascentis® Express HILIC (UV Detection)
HPLC Analysis of Paraquat and Diquat on Ascentis® Express HILIC (UV Detection)
Conditions
column Ascentis Express HILlC, 10 cm x 2.1 mm I.D., 2.7 µm particles (53939-U)
column temp. 60 °C
mobile phase [A] 200 mM ammonium TFA; [B] acetonitrile; (20:80, A:B)
flow rate 0.4 mL/min
sample 50 mg/L in mobile phase
injection 1 µL
detector UV, 257 nm (paraquat) and 308 nm (diquat)
Description
Categories Analytical Chromatography, Pesticides & Herbicides
Featured Industry Agriculture