HPLC Analysis of Peptides and Proteins on Proteomix® SCX-NP1.7, 5 cm x 4.6 mm I.D. Column
HPLC Analysis of Peptides and Proteins on Proteomix® SCX-NP1.7, 5 cm x 4.6 mm I.D. Column
Conditions
column Proteomix SCX-NP1.7, 5 cm x 4.6 mm I.D., 1.7 µm particles (Z777146)
column temp. ambient
mobile phase (A) 20 mM sodium phosphate, pH 6.0; (B) A + 1.0 M NaCl
gradient 0 to 75% B in 25 min; 15 min prewash
flow rate 0.75 ml/min
sample 1 mg/mL
injection 5 µl
detector UV, 214 nm
Description
Categories Analytical Chromatography, Proteins, Peptides
Featured Industry Life Science and Biopharma
Pharmaceutical (small molecule)
Legal Information Proteomix is a registered trademark of Sepax Technologies