HPLC Analysis of Peptides on Discovery® BIO PolyMA-SCX (Ion Exchange)
HPLC Analysis of Peptides on Discovery® BIO PolyMA-SCX (Ion Exchange)
Conditions
column Discovery BIO PolyMA-SCX, 5 cm x 4.6 mm I.D., 5 µm particles (59601-U)
column temp. 35 °C
mobile phase ([A] 5% acetonitrile in 20 mM ammonium carbonate, pH 3.5 with phosphoric acid; [B] 5% acetonitrile in 20 mM ammonium carbonate, 480 mM ammonium phosphate, pH 3.5 with phosphoric acid)
gradient 0 to 100% B in 24 min (linear)
flow rate 0.2 mL/min
sample RP Peptide Ionic Interactions Standard, p/n RPS-10020 (Alberta Peptide Institute)
injection 10 µL
detector UV, 210 nm
Description
Analysis Note Polymer particles are frequently used in bioseparations. Their advantages over inorganic particles, however, are often offset by generally lower efficiency. Discovery BIO PolyMA-SCX and PolyMA-WAX particles offer the benefits of polymeric particles, but have higher efficiency and better resolution than competitive particles. An example of the high efficiency of Discovery BIO PolyMA-SCX is shown in this Figure. The Discovery BIO PolyMA-SCX column gives efficient, well-resolved separation of a four-component peptide mixture.
Categories Analytical Chromatography, Peptides
Featured Industry Life Science and Biopharma
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC