HPLC Analysis of Peroxidase on TSKgel® G3000SWXL
HPLC Analysis of Peroxidase on TSKgel® G3000SWXL
Conditions
column TSKgel G3000SWXL, 30 cm x 7.8 mm I.D., 5 µm particles (808541)
column temp. 25 °C
mobile phase 0.3M sodium chloride/0.05M phosphate buffer, pH 7.0
flow rate 1 mL/min
injection 100 µL crude peroxidase preparation from Japanese horseradish (0.15mg)
detector UV, 220 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information TSKgel is a registered trademark of Tosoh Corporation