HPLC Analysis of Phenylephrine Enantiomers on Astec® CYCLOBOND® I 2000 AC
HPLC Analysis of Phenylephrine Enantiomers on Astec® CYCLOBOND® I 2000 AC
Conditions
column CYCLOBOND I 2000 AC, 25 cm x 4.6 mm I.D., 5 µm particles (20124AST)
column temp. 22 °C
mobile phase [A] 0.5% w/v sodium acetate, pH 5.5; [B] methanol; (90:10, A:B)
flow rate 0.5 mL/min
sample 1 mg/mL in methanol
injection 1 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Chiral Applications, Decongestants, Pharmaceutical (small molecule), All Pharmaceutical Applications
Featured Industry Pharmaceutical (small molecule)
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CYCLOBOND is a registered trademark of Sigma-Aldrich Co. LLC