HPLC Analysis of Propranolol Enantiomers on Astec® CHIROBIOTIC® T
HPLC Analysis of Propranolol Enantiomers on Astec® CHIROBIOTIC® T
Conditions
column CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 µm particles (12024AST)
column temp. 25 °C
mobile phase 15 mM ammonium formate in methanol
flow rate 1 mL/min
sample 1 mg/mL in methanol
injection 5 µL
detector UV, 220 nm
Description
Categories Analytical Chromatography, Chiral Applications, Antihypertensives, Pharmaceutical (small molecule), All Pharmaceutical Applications