HPLC Analysis of Proteins on TSKgel® CM-5PW
HPLC Analysis of Proteins on TSKgel® CM-5PW
Conditions
column TSKgel CM-5PW, 7.5 cm x 7.5 mm I.D., 10 µm particles (813068)
mobile phase [A] 0.1 M sodium phosphate, monbasic buffer, pH 7.0; [B] 0.5 M sodium chloride, A to B in 60 min
flow rate 1 mL/min
detector UV, 280 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information TSKgel is a registered trademark of Tosoh Corporation