HPLC Analysis of Proteins on TSKgel® Butyl-NPR
HPLC Analysis of Proteins on TSKgel® Butyl-NPR
Conditions
column TSKgel Butyl-NPR, 3.5 cm x 4.6 mm I.D., 2.5 µm particles (814947)
mobile phase [A] 0.1 M phosphate buffer, pH 7.0 plus ammonium sulfate, 2.3 M to 0 M in 12 min [B & C] = 0.1 M phosphate buffer, pH 7.0 plus ammonium sulfate, 1.8 M to 0 M in 60 min
flow rate 1 ml/min
detector UV, 280 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information TSKgel is a registered trademark of Tosoh Corporation