Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Proteins on Proteomix® SCX-NP1.7, Demonstration of Lot-to-Lot Reproducibility
HPLC Analysis of Proteins on Proteomix® SCX-NP1.7, Demonstration of Lot-to-Lot Reproducibility
Conditions
column Proteomix SCX-NP1.7, 5 cm x 4.6 mm I.D., 1.7 µm particles (Z777146)
column temp. ambient
mobile phase (A) 20 mM sodium phosphate buffer, pH 6.0, (B) A + 1.0 M NaCl
gradient 0 - 75% B in 25 min, 15 min prewash
flow rate 0.75 ml/min
sample 1 mg/mL each compound
injection 5 µl
detector UV, 214 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Pharmaceutical (small molecule)
Legal Information Proteomix is a registered trademark of Sepax Technologies