HPLC Analysis of Pseudoephedrine Enantiomers on Astec® CHIROBIOTIC® TAG
HPLC Analysis of Pseudoephedrine Enantiomers on Astec® CHIROBIOTIC® TAG
Conditions
column CHIROBIOTIC TAG, 25 cm x 4.6 mm I.D., 5 µm particles (14024AST)
column temp. 25 °C
mobile phase [A] methanol; [B] acetic acid; [C] triethylamine;(99.8:0.1:0.1, A:B:C)
flow rate 1 mL/min
sample 200 µg/mL in methanol
injection 10 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, StiµLants, Antihistamines, Decongestants