HPLC Analysis of Purines and Pyrimidines on SeQuant ZIC-HILIC
HPLC Analysis of Purines and Pyrimidines on SeQuant ZIC-HILIC
Conditions
column SeQuant ZIC-HILIC, PEEK, 150 x 2.1 mm, 5 µm, 200 Ã… (1.50454.0001)
mobile phase [A] acetonitrile; [B] ammonium formate; (80:20, A:B)
flow rate 0.1 mL/min
pressure (1.4 MPa)
injection 2 µL
detector 254 nm