HPLC Analysis of Pyroglutamic Acid Enantiomers on Astec® CHIROBIOTIC® T
HPLC Analysis of Pyroglutamic Acid Enantiomers on Astec® CHIROBIOTIC® T
Conditions
column CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 µm particles (12024AST)
column temp. 23 °C
mobile phase [A] acetonitrile; [B] methanol; [C] acetic acid; [D] triethylamine; (54.5:45:0.3:0.2, A:B:C:D)
flow rate 1 mL/min
sample 1 mg/mL in methanol
injection 5 µL
detector UV, 220 nm
Description
Categories Analytical Chromatography, Chiral Applications, Amino Acids, Life Science & Biopharma, All Biochemical Applications, All Amino Acids, Native Amino Acids