HPLC Analysis of α-Pyrrolidinovalerophenone (Flakka, PVP) Enantiomers on Astec® CHIROBIOTIC® V2
HPLC Analysis of α-Pyrrolidinovalerophenone (Flakka, PVP) Enantiomers on Astec® CHIROBIOTIC® V2
Conditions
column ASTEC CHIROBIOTIC V2 150X4.6MM (15023AST)
column temp. 20 °C
mobile phase [A ]Methanol [B] Water [C] Acetic Acid [D] Ammonium Hydroxide (25%); (95:5:0.1:0.02, A:B:C:D)
flow rate 1 mL/min
pressure 1100 psi (70 bar)
sample 100 µg/mL in methanol
injection 5 µL
detector UV, 251 nm
Description
Analysis Note Also known as a-PVP or O-2387, a-Pyrrolidinovalerophenone is a derivative of the synthetic cathinones MDPV and pyrovalerone. This illicit drug has stiµLant properties and has been found along with MDPV in several bath salt blends across the US, Europe, and Australia. Astec CHIROBIOTIC V2, based on vancomycin CSP, provided resolution of the enatiomers. CRMs from Cerillliant provided reliable identification and quantification.
Categories Analytical Chromatography, Drugs of Abuse
Featured Industry Forensics and Toxicology