HPLC Analysis of Salicylic Acid and Acetylsalicylic Acid (Aspirin) on Ascentis® Express OH5
HPLC Analysis of Salicylic Acid and Acetylsalicylic Acid (Aspirin) on Ascentis® Express OH5
Conditions
column Ascentis Express OH5, 10 cm x 3.0 mm I.D., 2.7 µm particles (53769-U)
column temp. 35 °C
mobile phase 2 mM ammonium acetate in 10:90 water:acetonitrile, pH to 7.0 with acetic acid, glacial
flow rate 0.5 mL/min
pressure 850 psi (59 bar)
sample 100 µg/mL salicylic acid, 200 µg/mL acetylsalicylic acid in methanol:acetonitrile (3:7)
injection 2 µL
detector UV, 235 nm
Description
Categories Analytical Chromatography, Analgesics