HPLC Analysis of Serotonin Synthetic Intermediates and Metabolites on Discovery® C18
HPLC Analysis of Serotonin Synthetic Intermediates and Metabolites on Discovery® C18
Conditions
column Discovery C18, 15 cm x 4.6 mm I.D., 5 µm particles (504955)
column temp. 35 °C
mobile phase [A] water (10 mM ammonium formate, pH 3.0 with formic acid): [B] acetonitrile; (95:5, A:B)
flow rate 1.0 mL/min
sample 50 µg/mL each
injection 10 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Clinical, hormones, Hormones
Featured Industry Clinical
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC