HPLC Analysis of Sinus Medications on Discovery® RP-AmideC16
HPLC Analysis of Sinus Medications on Discovery® RP-AmideC16
Conditions
column Discovery RP-AmideC16, 15 cm x 4.6 mm I.D., 5 µm particles (505013)
column temp. 30 °C
mobile phase [A] acetonitrile: [B] 50 mM KH2PO4, pH 3.0 (25:75, A:B)
flow rate 1 mL/min
injection 10 µL
detector UV, 220 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Analgesics
Featured Industry Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC