HPLC Analysis of Statins on Ascentis® RP-Amide
HPLC Analysis of Statins on Ascentis® RP-Amide
Conditions
column Ascentis RP-Amide, 15 cm x 4.6 mm I.D., 5 µm particles (565324-U)
column temp. 35 °C
mobile phase [A] water with 0.1% formic acid (34673): [B] methanol; (25:75, A:B)
flow rate 1.0 mL/min
sample 50 µg/mL in mobile phase
injection 10 µL
detector UV, 240 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Clinical, anticholesterolemics, Anticholesterolemics
Featured Industry Clinical
Pharmaceutical (small molecule)