HPLC Analysis of Steroids on Ascentis® Phenyl
HPLC Analysis of Steroids on Ascentis® Phenyl
Conditions
column Ascentis Phenyl, 15 cm x 4.6 mm I.D., 3 µm particles (581610-U)
column temp. 35 °C
mobile phase [A] water: [B] acetonitrile; (75:25, A:B)
flow rate 1.0 mL/min
sample 50 µg/mL in 85:15, water:methanol
injection 10 µL
detector UV, 240 nm
Description
Categories Analytical Chromatography, Clinical, steroids, Steroids