HPLC Analysis of Steroids on Ascentis® Express C18 (10 cm x 2.1 mm I.D.)
HPLC Analysis of Steroids on Ascentis® Express C18 (10 cm x 2.1 mm I.D.)
Conditions
column Ascentis Express C18, 10 cm x 2.1 mm I.D., 2.7 µm particles (53823-U)
column temp. 35 °C
mobile phase [A] water; [B] acetonitrile; (60:40, A:B)
flow rate 0.2 mL/min
pressure 2944 psi (203 bar)
sample 50 mg/L in 75:25, water:methanol
injection 2 µL
detector UV, 254 nm
Description
Analysis Note This application demonstrates the suitability of Ascentis Express C18 for the efficient separation of 5 steroids.
Categories Analytical Chromatography, Clinical, Steroids, steroids