HPLC Analysis of Strigol Enantiomers on Astec® CYCLOBOND® I 2000 RSP
HPLC Analysis of Strigol Enantiomers on Astec® CYCLOBOND® I 2000 RSP
Conditions
column CYCLOBOND I 2000 RSP, 25 cm x 4.6 mm I.D., 5 µm particles (20324AST)
column temp. 23 °C
mobile phase [A] 0.1% triethylamine, pH 4.1; [B] acetonitrile; (75:25, A:B)
flow rate 1 mL/min
sample 5 mg/mL in methanol
injection 5 µL
detector UV, 240 nm
Description
Categories Analytical Chromatography, Chiral Applications, Life Science & Biopharma, All Biochemical Applications, Other Biochemical Applications
Featured Industry Agriculture
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CYCLOBOND is a registered trademark of Sigma-Aldrich Co. LLC