HPLC Analysis of Sugars on SUPELCOGEL™ Ca
HPLC Analysis of Sugars on SUPELCOGEL™ Ca
Conditions
column SUPELCOGEL Ca, 30 cm x 7.8 mm I.D., 9 µm particles (59305-U)
column temp. 80 °C
mobile phase water
flow rate 1.5 mL/min
injection 10 µL, 2.5 mg/mL each analyte in water
detector RI
Description
Categories Analytical Chromatography, Carbohydrates
Featured Industry Food and Beverages
Legal Information SUPELCOGEL is a trademark of Sigma-Aldrich Co. LLC