HPLC Analysis of t-BOC-Alanine Enantiomers on Astec® CHIROBIOTIC® T
HPLC Analysis of t-BOC-Alanine Enantiomers on Astec® CHIROBIOTIC® T
Conditions
column CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 µm particles (12024AST)
column temp. 25 °C
mobile phase [A] 20 mM ammonium acetate, pH 6; [B] methanol; (90:10, A:B)
flow rate 1 mL/min
sample 5 mg/mL in methanol
injection 5 µL
detector UV, 230 nm
Description
Categories Analytical Chromatography, Chiral Applications, Amino Acids, Life Science & Biopharma, All Biochemical Applications, All Amino Acids, t-BOC-Amino Acids