HPLC Analysis of Thalidomide Enantiomers on Astec® CHIROBIOTIC® V
HPLC Analysis of Thalidomide Enantiomers on Astec® CHIROBIOTIC® V
Conditions
column CHIROBIOTIC V, 25 cm x 4.6 mm I.D., 5 µm particles (11024AST)
column temp. 22 °C
mobile phase [A] 18 mM ammonium nitrate, pH 5.5; [B] acetonitrile; (65:35, A:B)
flow rate 1 mL/min
sample 1 mg/mL in methanol
injection 20 µL
detector UV, 240 nm
Description
Categories Analytical Chromatography, Chiral Applications, Anxiolytics, Sedatives, Pharmaceutical (small molecule), Anxiolytics/Sedatives, All Pharmaceutical Applications